Tisk…

4-Bromo-3-methoxy-2-nitroaniline 95%

Dodavatel: Apollo Scientific
Varování

OR400689-1G OR400689-250MG
APOSOR400689-1GEA 11580 CZK
APOSOR400689-1G APOSOR400689-250MG
4-Bromo-3-methoxy-2-nitroaniline 95%
4-Bromo-3-methoxy-2-nitroaniline
Vzorec: C₇H₇BrN₂O₃
MW: 247,046 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28515361
CAS číslo: 1613265-20-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR