Tisk…

2-Bromo-4-iodo-6-nitrophenol

Dodavatel: Apollo Scientific
Varování

OR400673-1G OR400673-250MG
APOSOR400673-1GEA 13230 CZK
APOSOR400673-1G APOSOR400673-250MG
2-Bromo-4-iodo-6-nitrophenol
2-Bromo-4-iodo-6-nitrophenol
Vzorec: C₆H₃BrINO₃
MW: 343,901 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28515358
CAS číslo: 1936643-46-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR