Tisk…

4-Bromo-3-methylbenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1-Bromo-4-(bromomethyl)-2-methylbenzene

OR400670-1G OR400670-25G OR400670-5G
APOSOR400670-1GEA 1450 CZK
APOSOR400670-1G APOSOR400670-25G APOSOR400670-5G
4-Bromo-3-methylbenzyl bromide
4-Bromo-3-methylbenzyl bromide
Vzorec: C₈H₈Br₂
MW: 263.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909629
CAS číslo: 27561-51-9
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR