Tisk…

5-Bromo-3-chloroisatoic anhydride

Dodavatel: Apollo Scientific
Nebezpečí

OR400648-1G OR400648-250MG
APOSOR400648-1GEA 34390 CZK
APOSOR400648-1G APOSOR400648-250MG
5-Bromo-3-chloroisatoic anhydride
5-Bromo-3-chloroisatoic anhydride
Vzorec: C₈H₃BrClNO₃
MW: 276.471 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11899542
CAS číslo: 1006619-82-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR