4-Iodo-3-methoxybenzaldehyde 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400628-1G
OR400628-250MG
OR400628-5G
APOSOR400628-1GEA
3770
CZK
APOSOR400628-1G
APOSOR400628-250MG
APOSOR400628-5G
4-Iodo-3-methoxybenzaldehyde 95%
4-Iodo-3-methoxybenzaldehyde
Vzorec:
C₈H₇IO₂ Storage Temperature: Ambient |
MDL Number:
MFCD11847312 CAS číslo: 121404-83-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...