Tisk…

4-Bromo-2-nitrobenzyl alcohol

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-2-nitrobenzylalcohol

OR400605-25G OR400605-5G
APOSOR400605-25GEA 10520 CZK
APOSOR400605-25G APOSOR400605-5G
4-Bromo-2-nitrobenzyl alcohol
4-Bromo-2-nitrobenzyl alcohol
Vzorec: C₇H₆BrNO₃
MW: 232.031 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779493
CAS číslo: 22996-19-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR