Tisk…

2-Bromo-5-iodo-4-nitroanisole 95%

Dodavatel: Apollo Scientific
Varování

OR400544-1G OR400544-250MG OR400544-5G
APOSOR400544-1GEA 13490 CZK
APOSOR400544-1G APOSOR400544-250MG APOSOR400544-5G
2-Bromo-5-iodo-4-nitroanisole 95%
2-Bromo-5-iodo-4-nitroanisole
Vzorec: C₇H₅BrINO₃
MW: 357,928 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28348435
CAS číslo: 1361021-39-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR