Tisk…

2-Bromo-3-iodobenzaldehyde

Dodavatel: Apollo Scientific
Varování

OR400518-250MG OR400518-1G OR400518-5G
APOSOR400518-250MGEA 5390 CZK
APOSOR400518-250MG APOSOR400518-1G APOSOR400518-5G
2-Bromo-3-iodobenzaldehyde
2-Bromo-3-iodobenzaldehyde
Vzorec: C₇H₄BrIO
MW: 310.915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520407
CAS číslo: 1261850-39-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR