2-Chloro-6-methoxyphenol 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR400487-1G
OR400487-250MG
OR400487-5G
APOSOR400487-1GEA
6850
CZK
APOSOR400487-1G
APOSOR400487-250MG
APOSOR400487-5G
2-Chloro-6-methoxyphenol 95%
2-Chloro-6-methoxyphenol
Vzorec:
C₇H₇ClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD16998800 CAS číslo: 72403-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...