4-Methoxy-2-nitrobenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
OR400470-1G
OR400470-250MG
APOSOR400470-1GEA
12570
CZK
APOSOR400470-1G
APOSOR400470-250MG
4-Methoxy-2-nitrobenzyl alcohol
4-Methoxy-2-nitrobenzyl alcohol
Vzorec:
C₈H₉NO₄ MW: 183.161 g/mol Bod tání: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD05148477 CAS číslo: 22996-23-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...