4-Bromo-6-methoxy-1H-benzimidazole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
7-Bromo-5-methoxy-1H-benzo[d]imidazole
OR400429-1G
OR400429-250MG
OR400429-100MG
APOSOR400429-1GEA
16530
CZK
APOSOR400429-1G
APOSOR400429-250MG
APOSOR400429-100MG
4-Bromo-6-methoxy-1H-benzimidazole
4-Bromo-6-methoxy-1H-benzimidazole
Vzorec:
C₈H₇BrN₂O MW: 227,058 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22558067 CAS číslo: 1360954-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...