Tisk…

3-Bromo-4-iodobenzaldehyde

Dodavatel: Apollo Scientific
Varování

OR400418-1G OR400418-250MG OR400418-5G
APOSOR400418-1GEA 10750 CZK
APOSOR400418-1G APOSOR400418-250MG APOSOR400418-5G
3-Bromo-4-iodobenzaldehyde
3-Bromo-4-iodobenzaldehyde
Vzorec: C₇H₄BrIO
MW: 310,915 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779023
CAS číslo: 873387-82-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR