3-Bromo-4-iodobenzyl bromide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400405-1G
OR400405-250MG
OR400405-5G
APOSOR400405-1GEA
8270
CZK
APOSOR400405-1G
APOSOR400405-250MG
APOSOR400405-5G
3-Bromo-4-iodobenzyl bromide
3-Bromo-4-iodobenzyl bromide
Vzorec:
C₇H₅Br₂I Storage Temperature: Ambient |
MDL Number:
MFCD18391670 CAS číslo: 362529-52-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...