Tisk…

3-Bromo-4-iodobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

OR400404-1G OR400404-250MG
APOSOR400404-1GEA 14490 CZK
APOSOR400404-1G APOSOR400404-250MG
3-Bromo-4-iodobenzyl alcohol
3-Bromo-4-iodobenzyl alcohol
Vzorec: C₇H₆BrIO
MW: 312,93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09836677
CAS číslo: 249647-26-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR