Tisk…

Methyl 2-amino-4-cyanobenzoate

Dodavatel: Apollo Scientific

OR400400-1G OR400400-5G
APOSOR400400-1GEA 3970 CZK
APOSOR400400-1G APOSOR400400-5G
Methyl 2-amino-4-cyanobenzoate
Methyl 2-amino-4-cyanobenzoate
Vzorec: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20690800
CAS číslo: 159847-83-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR