Tisk…

4-Bromo-5-chlorobenzene-1,2-diamine 99%

Dodavatel: Apollo Scientific
Nebezpečí

OR400388-250MG OR400388-1G
APOSOR400388-250MGEA 1290 CZK
APOSOR400388-250MG APOSOR400388-1G
4-Bromo-5-chlorobenzene-1,2-diamine 99%
4-Bromo-5-chlorobenzene-1,2-diamine
Vzorec: C₆H₆BrClN₂
MW: 221,482 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16987754
CAS číslo: 75293-95-7
UN: 3259
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR