2-Amino-5-cyanobenzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
OR400360-1G
OR400360-5G
APOSOR400360-1GEA
3310
CZK
APOSOR400360-1G
APOSOR400360-5G
2-Amino-5-cyanobenzoic acid
2-Amino-5-cyanobenzoic acid
Vzorec:
C₈H₆N₂O₂ MW: 162.145 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD20485816 CAS číslo: 99767-45-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...