Methyl 2-amino-5-cyanobenzoate
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400352-1G
OR400352-250MG
APOSOR400352-1GEA
2940
CZK
APOSOR400352-1G
APOSOR400352-250MG
Methyl 2-amino-5-cyanobenzoate
Methyl 2-amino-5-cyanobenzoate
Vzorec:
C₉H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11041293 CAS číslo: 159847-81-1 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...