Tisk…

3-Chloro-5-methoxybenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

OR400328-1G OR400328-250MG OR400328-5G
APOSOR400328-1GEA 4630 CZK
APOSOR400328-1G APOSOR400328-250MG APOSOR400328-5G
3-Chloro-5-methoxybenzyl bromide
3-Chloro-5-methoxybenzyl bromide
Vzorec: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD08234654
CAS číslo: 885270-32-6
UN: 3261
ADR: 8,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR