3-Chloro-5-methoxybenzyl alcohol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
(3-Chloro-5-methoxyphenyl)methanol, 3-Chloro-5-(hydroxymethyl)anisole
OR400318-1G
OR400318-250MG
APOSOR400318-1GEA
4960
CZK
APOSOR400318-1G
APOSOR400318-250MG
3-Chloro-5-methoxybenzyl alcohol
3-Chloro-5-methoxybenzyl alcohol
Vzorec:
C₈H₉ClO₂ MW: 172.609 g/mol Bod varu: 130 °C (0.2 mm Hg) Bod tání: 37…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD08234652 CAS číslo: 82477-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...