4-Bromo-7-chloro-1H-benzimidazole 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR400259-1G
OR400259-250MG
APOSOR400259-1GEA
26060
CZK
APOSOR400259-1G
APOSOR400259-250MG
4-Bromo-7-chloro-1H-benzimidazole 95%
4-Bromo-7-chloro-1H-benzimidazole
Vzorec:
C₇H₄BrClN₂ Storage Temperature: Ambient |
MDL Number:
MFCD28040523 CAS číslo: 1360945-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...