Tisk…

4-Iodo-2-nitrophenol

Dodavatel: Apollo Scientific
Varování

OR400220-1G OR400220-5G OR400220-25G
APOSOR400220-1GEA 622 CZK
APOSOR400220-1G APOSOR400220-5G APOSOR400220-25G
4-Iodo-2-nitrophenol
4-Iodo-2-nitrophenol
Vzorec: C₆H₄INO₃
MW: 265,005 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466297
CAS číslo: 21784-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR