Tisk…

3-Methoxy-4-nitrobenzyl alcohol 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-Methoxy-4-nitrophenyl)methanol

OR400214-5G OR400214-10G
APOSOR400214-5GEA 2780 CZK
APOSOR400214-5G APOSOR400214-10G
3-Methoxy-4-nitrobenzyl alcohol 98%
3-Methoxy-4-nitrobenzyl alcohol
Vzorec: C₈H₉NO₄
MW: 183,161 g/mol
Bod tání: 95…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00007098
CAS číslo: 80866-88-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR