Tisk…

3-Bromo-5-methylbenzyl bromide

Dodavatel: Apollo Scientific
Nebezpečí

OR400207-1G OR400207-250MG OR400207-5G
APOSOR400207-1GEA 5590 CZK
APOSOR400207-1G APOSOR400207-250MG APOSOR400207-5G
3-Bromo-5-methylbenzyl bromide
3-Bromo-5-methylbenzyl bromide
Vzorec: C₈H₈Br₂
MW: 263,957 g/mol
Bod tání: 58…60 °C
Storage Temperature: Ambient
MDL Number: MFCD18204792
CAS číslo: 51719-69-8
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR