Tisk…

3-Bromo-5-methylphenylacetic acid 95%

Dodavatel: Apollo Scientific
Varování

OR400180-1G OR400180-250MG
APOSOR400180-1GEA 32240 CZK
APOSOR400180-1G APOSOR400180-250MG
3-Bromo-5-methylphenylacetic acid 95%
3-Bromo-5-methylphenylacetic acid
Vzorec: C₉H₉BrO₂
MW: 229,071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18392224
CAS číslo: 51719-66-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR