3-Chloro-2-nitroanisole
Dodavatel: Apollo Scientific
OR400163-1G
OR400163-5G
APOSOR400163-1GEA
2400
CZK
APOSOR400163-1G
APOSOR400163-5G
3-Chloro-2-nitroanisole
3-Chloro-2-nitroanisole
Vzorec:
C₇H₆ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD14455916 CAS číslo: 5472-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...