Tisk…

2-Iodo-6-methoxybenzonitrile

Dodavatel: Apollo Scientific
Varování

OR400161-1G OR400161-250MG OR400161-5G
APOSOR400161-1GEA 3940 CZK
APOSOR400161-1G APOSOR400161-250MG APOSOR400161-5G
2-Iodo-6-methoxybenzonitrile
2-Iodo-6-methoxybenzonitrile
Vzorec: C₈H₆INO
Storage Temperature: Ambient
MDL Number: MFCD18905184
CAS číslo: 66195-38-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR