2-Bromo-6-iodobenzaldehyde 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR400117-250MG
OR400117-1G
OR400117-5G
APOSOR400117-250MGEA
5690
CZK
APOSOR400117-250MG
APOSOR400117-1G
APOSOR400117-5G
2-Bromo-6-iodobenzaldehyde 95+%
2-Bromo-6-iodobenzaldehyde
Vzorec:
C₇H₄BrIO MW: 310,915 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520408 CAS číslo: 1261497-91-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...