Tisk…

Methyl 2-bromo-3-chlorobenzoate

Dodavatel: Apollo Scientific
Varování

OR400105-1G OR400105-5G OR400105-25G
APOSOR400105-1GEA 1740 CZK
APOSOR400105-1G APOSOR400105-5G APOSOR400105-25G
Methyl 2-bromo-3-chlorobenzoate
Methyl 2-bromo-3-chlorobenzoate
Vzorec: C₈H₆BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD18398719
CAS číslo: 1107627-14-4

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR