Tisk…

2-Bromo-3-chlorobenzyl bromide 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-1-(bromomethyl)-3-chlorobenzene

OR400096-1G OR400096-5G OR400096-250MG
APOSOR400096-1GEA 4960 CZK
APOSOR400096-1G APOSOR400096-5G APOSOR400096-250MG
2-Bromo-3-chlorobenzyl bromide 98%
2-Bromo-3-chlorobenzyl bromide
Vzorec: C₇H₅Br₂Cl
MW: 284,376 g/mol
Bod tání: 32,9…33.4 °C
Storage Temperature: Ambient
MDL Number: MFCD18390316
CAS číslo: 220453-60-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR