2-Bromo-3-methoxybenzyl bromide 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR400092-1G
OR400092-250MG
APOSOR400092-1GEA
22320
CZK
APOSOR400092-1G
APOSOR400092-250MG
2-Bromo-3-methoxybenzyl bromide 95%
2-Bromo-3-methoxybenzyl bromide
Vzorec:
C₈H₈Br₂O Bod tání: 60…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD18390184 CAS číslo: 128828-86-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...