Tisk…

4-Bromo-3-iodobenzyl alcohol

Dodavatel: Apollo Scientific
Varování

OR400084-250MG OR400084-1G
APOSOR400084-250MGEA 4960 CZK
APOSOR400084-250MG APOSOR400084-1G
4-Bromo-3-iodobenzyl alcohol
4-Bromo-3-iodobenzyl alcohol
Vzorec: C₇H₆BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD18391657
CAS číslo: 1261776-05-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR