Tisk…

2-Bromo-4-nitrobenzaldehyde

Dodavatel: Apollo Scientific
Varování

OR400069-1G OR400069-250MG OR400069-5G
APOSOR400069-1GEA 1870 CZK
APOSOR400069-1G APOSOR400069-250MG APOSOR400069-5G
2-Bromo-4-nitrobenzaldehyde
2-Bromo-4-nitrobenzaldehyde
Vzorec: C₇H₄BrNO₃
MW: 230,016 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08753661
CAS číslo: 5274-71-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR