Tisk…

2-Bromo-4-chlorobenzyl bromide 95%

Dodavatel: Apollo Scientific

OR400062-25G OR400062-5G
APOSOR400062-25GEA 9200 CZK
APOSOR400062-25G APOSOR400062-5G
2-Bromo-4-chlorobenzyl bromide 95%
2-Bromo-4-chlorobenzyl bromide
Vzorec: C₇H₅Br₂Cl
Storage Temperature: Ambient
MDL Number: MFCD12024978
CAS číslo: 33924-45-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR