Tisk…

4-Chloro-3-methoxyphenol 95%

Dodavatel: Apollo Scientific
Varování

OR400038-1G OR400038-250MG OR400038-5G
APOSOR400038-1GEA 2480 CZK
APOSOR400038-1G APOSOR400038-250MG APOSOR400038-5G
4-Chloro-3-methoxyphenol 95%
4-Chloro-3-methoxyphenol
Vzorec: C₇H₇ClO₂
MW: 158,582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16998742
CAS číslo: 18113-07-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR