4-Methoxyisatoic anhydride 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR400017-1G
OR400017-5G
OR400017-10G
APOSOR400017-1GEA
2730
CZK
APOSOR400017-1G
APOSOR400017-5G
APOSOR400017-10G
4-Methoxyisatoic anhydride 95%
4-Methoxyisatoic anhydride
Vzorec:
C₉H₇NO₄ MW: 193,156 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658459 CAS číslo: 128076-63-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...