3-Bromo-4-nitrophenylacetonitrile
Dodavatel: Apollo Scientific
OR399026-1G
OR399026-250MG
APOSOR399026-1GEA
11180
CZK
APOSOR399026-1G
APOSOR399026-250MG
3-Bromo-4-nitrophenylacetonitrile
3-Bromo-4-nitrophenylacetonitrile
Vzorec:
C₈H₅BrN₂O₂ MW: 241,041 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08275176 CAS číslo: 124840-60-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...