Tisk…

3-Bromo-2-nitrophenylacetonitrile

Dodavatel: Apollo Scientific

OR399022-1G OR399022-250MG
APOSOR399022-1GEA 11180 CZK
APOSOR399022-1G APOSOR399022-250MG
3-Bromo-2-nitrophenylacetonitrile
3-Bromo-2-nitrophenylacetonitrile
Vzorec: C₈H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18392324
CAS číslo: 185200-50-4
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR