Tisk…

3-Bromo-2-iodobenzoic acid

Dodavatel: Apollo Scientific
Varování

OR399017-1G OR399017-25G OR399017-5G
APOSOR399017-1GEA 2070 CZK
APOSOR399017-1G APOSOR399017-25G APOSOR399017-5G
3-Bromo-2-iodobenzoic acid
3-Bromo-2-iodobenzoic acid
Vzorec: C₇H₄BrIO₂
MW: 326,914 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15527220
CAS číslo: 503821-94-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR