Tisk…

2-Bromo-3-iodobenzoic acid 98%

Dodavatel: Apollo Scientific
Varování

OR399016-1G OR399016-5G
APOSOR399016-1GEA 1160 CZK
APOSOR399016-1G APOSOR399016-5G
2-Bromo-3-iodobenzoic acid 98%
2-Bromo-3-iodobenzoic acid
Vzorec: C₇H₄BrIO₂
MW: 326,914 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036150
CAS číslo: 855198-37-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR