Tisk…

3-Chloro-5-methoxyphenol

Dodavatel: Apollo Scientific

OR399015-5G
APOSOR399015-5GEA 4700 CZK
APOSOR399015-5G
3-Chloro-5-methoxyphenol
3-Chloro-5-methoxyphenol
Vzorec: C₇H₇ClO₂
MW: 158,582 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002260
CAS číslo: 65262-96-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR