Tisk…

3-Bromo-2-methoxyphenol

Dodavatel: Apollo Scientific

OR399009-1G OR399009-5G
APOSOR399009-1GEA 4140 CZK
APOSOR399009-1G APOSOR399009-5G
3-Bromo-2-methoxyphenol
3-Bromo-2-methoxyphenol
Vzorec: C₇H₇BrO₂
MW: 203,033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053054
CAS číslo: 88275-88-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR