3-Bromo-2-chlorophenol 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR399003-100G
OR399003-25G
APOSOR399003-100GEA
7770
CZK
APOSOR399003-100G
APOSOR399003-25G
3-Bromo-2-chlorophenol 98%
3-Bromo-2-chlorophenol
Vzorec:
C₆H₄BrClO MW: 207,452 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08166321 CAS číslo: 863870-87-5 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...