Tisk…

5,7-Dichloro-4-hydroxyquinoline-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR3929-1G OR3929-5G
APOSOR3929-1GEA 2480 CZK
APOSOR3929-1G APOSOR3929-5G
5,7-Dichloro-4-hydroxyquinoline-3-carboxylic acid
5,7-Dichloro-4-hydroxyquinoline-3-carboxylic acid
Vzorec: C₁₀H₅Cl₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD00219832
CAS číslo: 171850-30-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR