Tisk…

4-(3-Bromobenzyl)morpholine

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-[(3-Bromophenyl)methyl]morpholine

OR3762-5G OR3762-1G
APOSOR3762-5GEA 2730 CZK
APOSOR3762-5G APOSOR3762-1G
4-(3-Bromobenzyl)morpholine
4-(3-Bromobenzyl)morpholine
Vzorec: C₁₁H₁₄BrNO
MW: 256,139 g/mol
Hustota: 1,4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00979456
CAS číslo: 364793-82-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR