Tisk…

Benzo[d]oxazol-5-ol ≥98%

Dodavatel: Apollo Scientific

OR370097-100MG OR370097-1G OR370097-250MG
APOSOR370097-100MGEA 3150 CZK
APOSOR370097-100MG APOSOR370097-1G APOSOR370097-250MG
Benzo[d]oxazol-5-ol ≥98%
Benzo[d]oxazol-5-ol
Vzorec: C₇H₅NO₂
MW: 135,12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110248
CAS číslo: 180716-28-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR