Tisk…

N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline ≥98%

Dodavatel: Apollo Scientific

Synonyms: N-((1H-Imidazol-2-yl)methyl)-4-chloroaniline

OR370089-1G OR370089-5G
APOSOR370089-1GEA 5030 CZK
APOSOR370089-1G APOSOR370089-5G
N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline ≥98%
N-(1H-Imidazol-2-ylmethyl)-4-chloroaniline
Vzorec: C₁₀H₁₀ClN₃
MW: 207,659 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08668354
CAS číslo: 166096-16-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR