Tisk…

3-Bromopyridine-4-thiol ≥98%

Dodavatel: Apollo Scientific

OR370085-1G OR370085-5G
APOSOR370085-1GEA 8600 CZK
APOSOR370085-1G APOSOR370085-5G
3-Bromopyridine-4-thiol ≥98%
3-Bromopyridine-4-thiol
Vzorec: C₅H₄BrNS
MW: 190,061 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28337455
CAS číslo: 82264-72-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR