Chroman-6-ylboronic acid ≥97%
Dodavatel: Apollo Scientific
OR370080-1G
OR370080-250MG
APOSOR370080-1GEA
16770
CZK
APOSOR370080-1G
APOSOR370080-250MG
Chroman-6-ylboronic acid ≥97%
Chroman-6-ylboronic acid
Vzorec:
C₉H₁₁BO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD12779349 CAS číslo: 279261-84-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...