4-Chloro-1,8-dimethyl-imidazo[1,2-a]quinoxaline ≥98%
Dodavatel: Apollo Scientific
OR370073-100MG
OR370073-250MG
APOSOR370073-100MGEA
13060
CZK
APOSOR370073-100MG
APOSOR370073-250MG
4-Chloro-1,8-dimethyl-imidazo[1,2-a]quinoxaline ≥98%
4-Chloro-1,8-dimethyl-imidazo[1,2-a]quinoxaline
Vzorec:
C₁₂H₁₀ClN₃ MW: 231,681 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15528883 CAS číslo: 445430-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...